Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYAT1 | NM_001122671.1 | NP_001116143.1 | ||
| NM_001122672.1 | NP_001116144.1 | |||
| NM_001287390.1 | NP_001274319.1 | |||
| NM_004059.4 | NP_004050.3 | |||
| NR_109829.1 | ||||
| XM_006717311.3 | XP_006717374.1 | |||
| XM_011519165.2 | XP_011517467.1 | |||
| XM_011519167.2 | XP_011517469.2 | |||
| XM_011519169.1 | XP_011517471.1 | |||
| XM_017015260.1 | XP_016870749.1 | |||
| XM_017015261.1 | XP_016870750.1 | |||
| XM_017015264.1 | XP_016870753.1 | |||
| XM_017015266.1 | XP_016870755.1 | |||
| XM_017015267.1 | XP_016870756.1 | |||
| XM_017015268.1 | XP_016870757.1 | |||
| AK291178.1 | ||||
| AU123740.1 | ||||
| DA381449.1 |
Target Gene Details
Entrez Gene ID: | 56262 |
Gene Name: | leucine rich repeat containing 8 family member A |
Gene Aliases: |
AGM5, LRRC8, SWELL1 |
Location: |
Chr.9:128882112-128918042 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC8A | NM_001127245.1 | NP_001120717.1 | ||
| NM_019594.3 | NP_062540.2 | |||
| DA243590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621819 | Chr.9:128877077 - 128887549 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map