Assay Details
Target Gene Details
Entrez Gene ID: | 5557 |
Gene Name: | primase (DNA) subunit 1 |
Gene Aliases: |
p49 |
Location: |
Chr.12:56731580-56752362 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRIM1 | NM_000946.2 | NP_000937.1 | ||
| AK223391.1 | BAD97111.1 | |||
| AK314866.1 | ||||
| BC005266.1 | AAH05266.1 | |||
| BU167590.1 | ||||
| CD686660.1 | ||||
| X74330.1 | CAA52377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map