Assay Details
Target Gene Details
Entrez Gene ID: | 112950 |
Gene Name: | mediator complex subunit 8 |
Gene Aliases: |
ARC32 |
Location: |
Chr.1:43383908-43389812 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED8 | NM_001001653.2 | 7 | 1272 | NP_001001653.1 |
| NM_052877.4 | NP_443109.2 | |||
| NM_201542.4 | 7 | 1229 | NP_963836.2 | |
| BC010543.1 | ||||
| CB161514.1 | 1 | 129 | ||
| CB162354.1 | 1 | 129 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map