Assay Details
Target Gene Details
Entrez Gene ID: | 55709 |
Gene Name: | kelch repeat and BTB domain containing 4 |
Gene Aliases: |
BKLHD4, HSPC252 |
Location: |
Chr.11:47572197-47579015 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
| nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion |
|

Genomic Map