Assay Details
Target Gene Details
Entrez Gene ID: | 340547 |
Gene Name: | V-set and immunoglobulin domain containing 1 |
Gene Aliases: |
1700062D20Rik, GPA34, dJ889N15.1 |
Location: |
Chr.X:108018886-108079184 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG1 | NM_001170553.1 | NP_001164024.1 | ||
| NM_182607.4 | NP_872413.1 | |||
| XM_005262127.4 | XP_005262184.1 | |||
| XM_011530936.2 | XP_011529238.1 | |||
| AK292472.1 | ||||
| AK301311.1 | ||||
| AK301783.1 | ||||
| AK315846.1 | ||||
| BC043216.2 | AAH43216.1 | |||
| BX640913.1 | CAE45954.1 | |||
| BX648658.1 | CAH56142.1 |
More Information
Set Membership: |
|

Genomic Map