Assay Details
Target Gene Details
Entrez Gene ID: | 2885 |
Gene Name: | growth factor receptor bound protein 2 |
Gene Aliases: |
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2 |
Location: |
Chr.17:75318076-75405709 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRB2 | NM_002086.4 | NP_002077.1 | ||
| NM_203506.2 | NP_987102.1 | |||
| AF498925.1 | AAM21073.1 | |||
| AK091010.1 | ||||
| AK315565.1 | ||||
| BC000631.2 | AAH00631.1 | |||
| BI459414.1 | ||||
| CR450363.1 | CAG29359.1 | |||
| CR541942.1 | CAG46740.1 | |||
| DA458680.1 | ||||
| L29511.1 | AAC37549.1 | |||
| M96995.1 | AAA58448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833541 | Chr.17:75297204 - 75460023 on Build GRCh38 | Loss |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map