Assay Details
Target Gene Details
Entrez Gene ID: | 6464 |
Gene Name: | SHC adaptor protein 1 |
Gene Aliases: |
SHC, SHCA |
Location: |
Chr.1:154962298-154974492 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map