Assay Details
Target Gene Details
Entrez Gene ID: | 144568 |
Gene Name: | alpha-2-macroglobulin like 1 |
Gene Aliases: |
CPAMD9 |
Location: |
Chr.12:8822472-8887202 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A2ML1 | NM_144670.5 | NP_653271.2 | ||
| XM_011520566.2 | XP_011518868.1 | |||
| XM_011520567.2 | XP_011518869.1 | |||
| XM_017018868.1 | XP_016874357.1 | |||
| XM_017018869.1 | XP_016874358.1 | |||
| XM_017018870.1 | XP_016874359.1 | |||
| AK122624.1 | BAC85494.1 | |||
| AK290355.1 | ||||
| AL832139.1 | ||||
| BX648231.1 | ||||
| DA436610.1 | ||||
| DA456454.1 | ||||
| DA461408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map