Assay Details
Target Gene Details
Entrez Gene ID: | 79187 |
Gene Name: | fibronectin type III and SPRY domain containing 1 |
Gene Aliases: |
GLFND, MIR1 |
Location: |
Chr.19:4304594-4323846 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSD1 | NM_024333.2 | NP_077309.1 | ||
| XM_005259644.2 | XP_005259701.1 | |||
| AF316829.1 | AAK26747.1 | |||
| AK021750.1 | BAB13885.1 | |||
| AK315661.1 | ||||
| AY032617.1 | AAK51145.1 | |||
| BC003124.1 | AAH03124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578442 | Chr.19:4236999 - 4312106 on Build GRCh38 | Loss |
|
| nsv833717 | Chr.19:4204757 - 4368519 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map