Assay Details
Target Gene Details
Entrez Gene ID: | 54931 |
Gene Name: | tRNA methyltransferase 10C, mitochondrial RNase P subunit |
Gene Aliases: |
COXPD30, HNYA, MRPP1, RG9MTD1 |
Location: |
Chr.3:101561836-101566446 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT10C | NM_017819.3 | 2 | 778 | NP_060289.2 |
| AF226052.1 | 2 | 746 | AAF86952.1 | |
| AK000439.1 | 2 | 745 | BAA91166.1 | |
| AY561707.1 | 1 | 580 | AAS66980.1 | |
| BC035967.1 | 2 | 752 | AAH35967.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508944 | Chr.3:101528526 - 101575525 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map