Assay Details
Target Gene Details
Entrez Gene ID: | 5275 |
Gene Name: | serpin family B member 13 |
Gene Aliases: |
HSHUR7SEQ, HUR7, PI13, headpin |
Location: |
Chr.18:63587206-63599199 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB13 | NM_001307923.1 | NP_001294852.1 | ||
| NM_012397.3 | NP_036529.1 | |||
| XM_005266707.3 | XP_005266764.1 | |||
| XM_011526029.2 | XP_011524331.1 | |||
| AF169949.1 | AAD55765.1 | |||
| AJ001696.2 | CAA04935.2 | |||
| AJ001697.2 | CAA04936.2 | |||
| AJ001698.1 | CAA04937.1 | |||
| AK290323.1 | ||||
| AK296797.1 | ||||
| AK298049.1 | ||||
| AK303867.1 | ||||
| AK303979.1 | ||||
| AK313486.1 | ||||
| BC101821.1 | AAI01822.1 | |||
| BC143402.1 | ||||
| X98307.1 | 1 | 117 | CAA66951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map