Assay Details
Target Gene Details
Entrez Gene ID: | 58191 |
Gene Name: | C-X-C motif chemokine ligand 16 |
Gene Aliases: |
CXCLG16, SR-PSOX, SRPSOX |
Location: |
Chr.17:4733529-4739928 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL16 | NM_001100812.1 | NP_001094282.1 | ||
| NM_022059.3 | NP_071342.2 | |||
| AF275260.1 | AAG31750.1 | |||
| AF301016.1 | AAG34365.1 | |||
| AF337812.1 | AAK38275.1 | |||
| AK027389.1 | BAB55078.1 | |||
| AK292114.1 | ||||
| AK315017.1 | ||||
| AY358909.1 | AAQ89268.1 | |||
| BC017588.1 | AAH17588.1 | |||
| BC044930.1 | AAH44930.1 | |||
| CA307348.1 | ||||
| JC209418.1 | ||||
| JC209420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
| nsv1952 | Chr.17:4726245 - 4748889 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map