Assay Details
Target Gene Details
Entrez Gene ID: | 2305 |
Gene Name: | forkhead box M1 |
Gene Aliases: |
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT |
Location: |
Chr.12:2857681-2877155 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXM1 | NM_001243088.1 | NP_001230017.1 | ||
| NM_001243089.1 | NP_001230018.1 | |||
| NM_021953.3 | NP_068772.2 | |||
| NM_202002.2 | NP_973731.1 | |||
| NM_202003.2 | NP_973732.1 | |||
| XM_005253676.3 | XP_005253733.1 | |||
| XM_011520930.2 | XP_011519232.1 | |||
| XM_011520931.2 | XP_011519233.1 | |||
| XM_011520932.1 | XP_011519234.1 | |||
| XM_011520933.1 | XP_011519235.1 | |||
| XM_011520934.2 | XP_011519236.1 | |||
| XM_011520935.1 | XP_011519237.1 | |||
| BQ229497.1 | ||||
| DB137846.1 |
Target Gene Details
Entrez Gene ID: | 83695 |
Gene Name: | RAD9-HUS1-RAD1 interacting nuclear orphan 1 |
Gene Aliases: |
C12orf32, HKMT1188, RHINO |
Location: |
Chr.12:2876258-2889525 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHNO1 | NM_001252499.2 | NP_001239428.1 | ||
| NM_001252500.2 | NP_001239429.1 | |||
| NM_001257097.1 | NP_001244026.1 | |||
| NM_001257098.1 | NP_001244027.1 | |||
| NR_027365.2 | ||||
| NR_046432.1 | ||||
| NR_046433.1 | ||||
| BM847461.1 | ||||
| DA679606.1 | ||||
| DA965227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map