Assay Details
Target Gene Details
Entrez Gene ID: | 6527 |
Gene Name: | solute carrier family 5 member 4 |
Gene Aliases: |
DJ90G24.4, SAAT1, SGLT3 |
Location: |
Chr.22:32218423-32355057 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A4 | NM_014227.2 | NP_055042.1 | ||
| XM_006724308.3 | XP_006724371.1 | |||
| XM_011530342.2 | XP_011528644.1 | |||
| XM_011530343.2 | XP_011528645.1 | |||
| XM_011530344.2 | XP_011528646.1 | |||
| XM_017028920.1 | XP_016884409.1 | |||
| AJ133127.1 | CAB81772.1 | |||
| U41897.1 | AAB61732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
|
| esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
|
| nsv588895 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| nsv834178 | Chr.22:32094462 - 32281534 on Build GRCh38 | Loss |
|
| esv3893475 | Chr.22:32228077 - 32260035 on Build GRCh38 | Gain |
|
| esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map