Assay Details
Target Gene Details
Entrez Gene ID: | 10036 |
Gene Name: | chromatin assembly factor 1 subunit A |
Gene Aliases: |
CAF-1, CAF1, CAF1B, CAF1P150, P150 |
Location: |
Chr.19:4402596-4448322 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAF1A | NM_005483.2 | 3 | 458 | NP_005474.2 |
| XM_011527605.2 | 3 | 525 | XP_011525907.1 | |
| XM_011527606.2 | 3 | 525 | XP_011525908.1 | |
| XM_011527607.2 | 3 | 525 | XP_011525909.1 | |
| BC052620.1 | 3 | 457 | AAH52620.1 | |
| BC067093.1 | 3 | 459 | AAH67093.1 | |
| BQ889365.1 | 3 | 458 | ||
| U20979.1 | 3 | 344 | AAA76736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833720 | Chr.19:4367721 - 4540556 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map