Assay Details
Target Gene Details
Entrez Gene ID: | 1241 |
Gene Name: | leukotriene B4 receptor |
Gene Aliases: |
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7 |
Location: |
Chr.14:24311499-24318036 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTB4R | NM_001143919.2 | NP_001137391.1 | ||
| D89079.1 | BAA20424.1 | |||
| U41070.1 | AAC50628.1 |
Target Gene Details
Entrez Gene ID: | 56413 |
Gene Name: | leukotriene B4 receptor 2 |
Gene Aliases: |
BLT2, BLTR2, JULF2, KPG_004, LTB4-R 2, LTB4-R2, NOP9 |
Location: |
Chr.14:24310151-24312053 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTB4R2 | NM_001164692.2 | 2 | 1003 | NP_001158164.1 |
| NM_019839.4 | 2 | 947 | NP_062813.2 | |
| AB029892.1 | 1 | 832 | BAA99553.1 | |
| AB032558.1 | 2 | 913 | BAD83590.1 | |
| AB044402.1 | 1 | 832 | BAA99539.1 | |
| AF277230.1 | 1 | 2543 | AAF81261.1 | |
| AF308571.1 | 1 | 832 | AAG40857.1 | |
| AK304169.1 | 2 | 939 | ||
| BC119674.2 | ||||
| BC127643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564058 | Chr.14:24302079 - 24317770 on Build GRCh38 | Loss |
|
| nsv564060 | Chr.14:24307195 - 24340589 on Build GRCh38 | Loss |
|
| esv2669637 | Chr.14:24274158 - 24318169 on Build GRCh38 | Deletion |
|
| esv3633804 | Chr.14:24274134 - 24318147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map