Assay Details
Target Gene Details
Entrez Gene ID: | 11278 |
Gene Name: | Kruppel like factor 12 |
Gene Aliases: |
AP-2rep, AP2REP, HSPC122 |
Location: |
Chr.13:73686012-74168295 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF12 | NM_007249.4 | 8 | 7605 | NP_009180.3 |
| XM_005266251.3 | 8 | 7714 | XP_005266308.1 | |
| XM_011534907.2 | 7 | 7428 | XP_011533209.1 | |
| XM_011534909.2 | 8 | 7561 | XP_011533211.1 | |
| XM_011534910.2 | 7 | 7392 | XP_011533212.1 | |
| XM_011534911.2 | 7 | 7470 | XP_011533213.1 | |
| XM_011534912.2 | 7 | 7541 | XP_011533214.1 | |
| XM_017020384.1 | 7 | 7372 | XP_016875873.1 | |
| XM_017020385.1 | 8 | 7645 | XP_016875874.1 | |
| AF330041.1 | 7 | 7391 | AAL37478.1 | |
| AK094419.1 | 1 | 245 | ||
| AK126412.1 | 1 | 819 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562343 | Chr.13:73228534 - 73782546 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map