Assay Details
Target Gene Details
Entrez Gene ID: | 5458 |
Gene Name: | POU class 4 homeobox 2 |
Gene Aliases: |
BRN3.2, BRN3B, Brn-3b |
Location: |
Chr.4:146638893-146642471 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU4F2 | NM_004575.2 | 2 | 2104 | NP_004566.2 |
| BC136344.1 | 2 | 2011 | ||
| BC136345.1 | 2 | 2011 | ||
| EU439706.1 | 2 | 1840 | ||
| U06233.1 | 2 | 2071 | AAA16509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822768 | Chr.4:146635834 - 146641502 on Build GRCh38 | Gain |
|
| esv3894049 | Chr.4:146348182 - 146767074 on Build GRCh38 | Gain |
|
| nsv595651 | Chr.4:146195712 - 146847054 on Build GRCh38 | Gain |
|
| nsv830107 | Chr.4:146531282 - 146714568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map