Assay Details
Target Gene Details
Entrez Gene ID: | 5793 |
Gene Name: | protein tyrosine phosphatase, receptor type G |
Gene Aliases: |
HPTPG, PTPG, R-PTP-GAMMA, RPTPG |
Location: |
Chr.3:61561569-62297609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRG | NM_002841.3 | NP_002832.3 | ||
| XM_005265353.3 | XP_005265410.1 | |||
| XM_017006961.1 | XP_016862450.1 | |||
| XM_017006962.1 | XP_016862451.1 | |||
| XM_017006963.1 | XP_016862452.1 | |||
| XM_017006964.1 | XP_016862453.1 | |||
| AB209871.1 | BAD93108.1 | |||
| AK296327.1 | ||||
| BC140904.1 | ||||
| BC144110.1 | ||||
| CR749269.1 | CAH18125.1 | |||
| L09247.1 | AAA60224.1 | |||
| X54132.1 | CAA38067.1 |
Target Gene Details
Entrez Gene ID: | 100506994 |
Gene Name: | PTPRG antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:62261819-62318947 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRG-AS1 | NR_038281.1 | 5 | 1663 | |
| NR_038282.1 | 5 | 1649 | ||
| NR_038283.1 | 4 | 1567 | ||
| AK022901.1 | 3 | 1419 | ||
| AK094634.1 | 4 | 1543 |
More Information
Set Membership: |
|

Genomic Map