Assay Details
Target Gene Details
Entrez Gene ID: | 255189 |
Gene Name: | phospholipase A2 group IVF |
Gene Aliases: |
PLA2G4FZ |
Location: |
Chr.15:42141134-42156641 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4F | NM_213600.3 | 16 | 1827 | NP_998765.3 |
| NR_033151.1 | 15 | 1754 | ||
| AK122623.1 | 16 | 1798 | ||
| AK172836.1 | BAD18801.1 | |||
| AK304368.1 | 16 | 1809 | ||
| BC035335.1 | 15 | 1724 | AAH35335.1 | |
| BC131531.1 | 16 | 1739 | ||
| BC142682.1 | 16 | 1798 | ||
| BC146648.1 | 16 | 1798 | ||
| CR749451.1 | 9 | 3900 | CAH18288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv952614 | Chr.15:42139503 - 42164202 on Build GRCh38 | Deletion |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map