Assay Details
Target Gene Details
Entrez Gene ID: | 7775 |
Gene Name: | zinc finger protein 232 |
Gene Aliases: |
ZSCAN11 |
Location: |
Chr.17:5105734-5123116 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF232 | NM_001320952.1 | 4 | 1634 | NP_001307881.1 |
| NM_001320953.1 | 6 | 1948 | NP_001307882.1 | |
| NM_001320954.1 | 6 | 1893 | NP_001307883.1 | |
| NM_001320955.1 | 4 | 1577 | NP_001307884.1 | |
| NM_014519.3 | 4 | 1661 | NP_055334.2 | |
| XM_011524006.2 | 5 | 1327 | XP_011522308.1 | |
| XM_017025021.1 | 5 | 1878 | XP_016880510.1 | |
| XM_017025022.1 | 5 | 1244 | XP_016880511.1 | |
| XM_017025023.1 | 5 | 1218 | XP_016880512.1 | |
| AK057506.1 | 3 | 1692 | ||
| AK225622.1 | 4 | 1191 | ||
| AK298190.1 | ||||
| BC016039.1 | 4 | 1246 | AAH16039.2 | |
| BQ184519.1 | 1 | 231 | ||
| Y15067.1 | 4 | 1253 | CAB95963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map