Assay Details
Target Gene Details
Entrez Gene ID: | 23646 |
Gene Name: | phospholipase D family member 3 |
Gene Aliases: |
AD19, HU-K4, HUK4 |
Location: |
Chr.19:40348395-40378490 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD3 | NM_001031696.3 | NP_001026866.1 | ||
| NM_001291311.1 | NP_001278240.1 | |||
| NM_012268.3 | NP_036400.2 | |||
| XM_005258704.1 | XP_005258761.1 | |||
| XM_005258707.4 | XP_005258764.1 | |||
| XM_005258708.3 | XP_005258765.1 | |||
| XM_005258709.4 | XP_005258766.1 | |||
| XM_005258710.4 | XP_005258767.1 | |||
| XM_006723122.1 | XP_006723185.1 | |||
| XM_011526692.1 | XP_011524994.1 | |||
| XM_011526693.1 | XP_011524995.1 | |||
| XM_017026546.1 | XP_016882035.1 | |||
| XM_017026547.1 | XP_016882036.1 | |||
| XM_017026548.1 | XP_016882037.1 | |||
| XM_017026549.1 | XP_016882038.1 | |||
| AK293690.1 | ||||
| AY927565.1 | 2 | 261 | ||
| BC000553.1 | AAH00553.2 | |||
| BC036327.1 | AAH36327.1 | |||
| BC096820.1 | AAH96820.1 | |||
| BP304863.1 | ||||
| U60644.1 | AAB16799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833831 | Chr.19:40290833 - 40454958 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map