Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | NP_001252541.1 | ||
| NM_016284.4 | NP_057368.3 | |||
| NR_049763.1 | ||||
| AB023224.2 | BAA76851.2 | |||
| AF110778.1 | AAF14861.1 | |||
| BM023419.1 | ||||
| CR749237.1 | CAH18093.1 |
Target Gene Details
Entrez Gene ID: | 79918 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.16:58515479-58521961 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD6 | NM_001160305.2 | 8 | 2427 | NP_001153777.1 |
| NM_024860.3 | 9 | 2355 | NP_079136.2 | |
| NR_134583.1 | 9 | 2386 | ||
| AK127136.1 | 5 | 3832 | BAC86845.1 |
More Information
Set Membership: |
|

Genomic Map