Assay Details
Target Gene Details
Entrez Gene ID: | 87 |
Gene Name: | actinin alpha 1 |
Gene Aliases: |
BDPLT15 |
Location: |
Chr.14:68874123-68979366 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
More Information
Set Membership: |
|

Genomic Map