Assay Details
Target Gene Details
Entrez Gene ID: | 57379 |
Gene Name: | activation-induced cytidine deaminase |
Gene Aliases: |
AID, ARP2, CDA2, HEL-S-284, HIGM2 |
Location: |
Chr.12:8602166-8612970 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AICDA | NM_020661.2 | NP_065712.1 | ||
| XM_011520772.2 | XP_011519074.1 | |||
| XM_011520773.2 | XP_011519075.1 | |||
| AB040431.1 | BAB12721.1 | |||
| AF529815.1 | AAM95402.1 | |||
| AF529816.1 | AAM95403.1 | |||
| AF529817.1 | AAM95404.1 | |||
| AF529818.1 | AAM95405.1 | |||
| AF529819.1 | AAM95406.1 | |||
| AF529820.1 | AAM95407.1 | |||
| AF529821.1 | AAM95408.1 | |||
| AF529822.1 | AAM95409.1 | |||
| AF529823.1 | AAM95410.1 | |||
| AF529824.1 | AAM95411.1 | |||
| AF529825.1 | AAM95412.1 | |||
| AF529826.1 | AAM95413.1 | |||
| AF529827.1 | AAM95414.1 | |||
| AY534975.1 | AAS90604.1 | |||
| AY536516.1 | AAS92920.1 | |||
| AY536517.1 | AAS92921.1 | |||
| AY541058.1 | AAS98257.1 | |||
| BC006296.2 | AAH06296.1 | |||
| BT007402.1 | AAP36066.1 | |||
| BX402063.2 | ||||
| DQ431660.1 | ||||
| EU794587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss |
|
| dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map