Assay Details
Target Gene Details
Entrez Gene ID: | 2150 |
Gene Name: | F2R like trypsin receptor 1 |
Gene Aliases: |
GPR11, PAR2 |
Location: |
Chr.5:76819008-76835315 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2RL1 | XM_017009223.1 | 2 | 906 | XP_016864712.1 |
| AY336105.1 | 2 | 928 | AAP97012.1 | |
| BC012453.1 | 2 | 939 | AAH12453.1 | |
| BC018130.1 | 2 | 1001 | AAH18130.1 | |
| BT009856.1 | 2 | 847 | AAP88858.1 | |
| U34038.1 | 2 | 994 | AAB47871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5703n100 | Chr.5:76783895 - 76838952 on Build GRCh38 | Gain |
|
| esv3605474 | Chr.5:76789709 - 76840353 on Build GRCh38 | Gain |
|
| esv994503 | Chr.5:76833405 - 76848357 on Build GRCh38 | Deletion |
|
| esv2763908 | Chr.5:76787569 - 76838952 on Build GRCh38 | Gain |
|
| dgv9885n54 | Chr.5:76790772 - 76834288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map