Assay Details
Target Gene Details
Entrez Gene ID: | 9950 |
Gene Name: | golgin A5 |
Gene Aliases: |
GOLIM5, RFG5, ret-II |
Location: |
Chr.14:92794231-92839963 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA5 | NM_005113.3 | 2 | 752 | NP_005104.3 |
| XM_011537420.2 | 2 | 752 | XP_011535722.1 | |
| AF085199.1 | 2 | 627 | AAD09753.1 | |
| AJ132949.1 | 1 | 496 | CAB36967.1 | |
| AK293077.1 | 2 | 655 | ||
| AK303789.1 | ||||
| AY644768.1 | 1 | 532 | AAV85456.1 | |
| BC023021.1 | 2 | 660 | AAH23021.1 |
More Information
Set Membership: |
|

Genomic Map