Assay Details
Target Gene Details
Entrez Gene ID: | 6942 |
Gene Name: | transcription factor 20 |
Gene Aliases: |
AR1, SPBP, TCF-20 |
Location: |
Chr.22:42160013-42283927 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF20 | NM_005650.3 | 6 | 7005 | NP_005641.1 |
| NM_181492.2 | 4 | 6776 | NP_852469.1 | |
| XM_005261722.3 | 6 | 8208 | XP_005261779.1 | |
| XM_006724313.3 | 6 | 7189 | XP_006724376.1 | |
| XM_011530353.2 | 6 | 7156 | XP_011528655.1 | |
| XM_011530354.2 | 5 | 7061 | XP_011528656.1 | |
| AB006630.1 | 5 | 6137 | BAA22961.1 | |
| BU732753.1 | 2 | 424 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3643 | Chr.22:42134472 - 42197574 on Build GRCh38 | Insertion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map