Assay Details
Target Gene Details
Entrez Gene ID: | 63946 |
Gene Name: | DMRT like family C2 |
Gene Aliases: |
- |
Location: |
Chr.19:41844743-41852330 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMRTC2 | NM_001040283.2 | 6 | 771 | NP_001035373.1 |
| XM_011527214.1 | 6 | 783 | XP_011525516.1 | |
| XM_011527215.1 | 6 | 775 | XP_011525517.1 | |
| XM_017027123.1 | 6 | 788 | XP_016882612.1 | |
| XM_017027124.1 | 6 | 790 | XP_016882613.1 | |
| XM_017027125.1 | 6 | 793 | XP_016882614.1 | |
| XM_017027126.1 | 6 | 883 | XP_016882615.1 | |
| XM_017027127.1 | 6 | 748 | XP_016882616.1 | |
| XM_017027128.1 | 6 | 953 | XP_016882617.1 | |
| XM_017027129.1 | 6 | 784 | XP_016882618.1 | |
| AJ291669.1 | 5 | 587 | CAC40652.1 | |
| AK057404.1 | 6 | 775 | BAB71473.1 | |
| AK301822.1 | 6 | 774 | ||
| BC029202.1 | 6 | 737 | AAH29202.1 | |
| BC039266.1 | 6 | 739 | AAH39266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map