Assay Details
Target Gene Details
Entrez Gene ID: | 2152 |
Gene Name: | coagulation factor III, tissue factor |
Gene Aliases: |
CD142, TF, TFA |
Location: |
Chr.1:94529176-94541857 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F3 | NM_001178096.1 | NP_001171567.1 | ||
| NM_001993.4 | NP_001984.1 | |||
| AF487337.1 | AAO61150.1 | |||
| AF497569.1 | AAO61151.1 | |||
| AF497570.1 | AAO61152.1 | |||
| AK310918.1 | 3 | 800 | ||
| AK312638.1 | ||||
| AY940729.1 | ||||
| AY940730.1 | ||||
| AY940731.1 | ||||
| BC011029.1 | AAH11029.1 | |||
| BT019808.1 | AAV38611.1 | |||
| CR541792.1 | CAG46591.1 | |||
| DB203987.1 | ||||
| DB499288.1 | ||||
| J02931.1 | AAA61150.1 | |||
| M16553.1 | AAA61151.1 | |||
| M27436.1 | AAA36734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1899 | Chr.1:94534821 - 94564202 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map