Assay Details
Target Gene Details
Entrez Gene ID: | 9441 |
Gene Name: | mediator complex subunit 26 |
Gene Aliases: |
CRSP7, CRSP70 |
Location: |
Chr.19:16574907-16628204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED26 | NM_004831.3 | NP_004822.2 | ||
| AF104253.1 | AAD12722.1 | |||
| AK128435.1 | 1 | 362 | ||
| AK313778.1 | ||||
| BC110430.1 | ||||
| BC127215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510757 | Chr.19:16559921 - 16632309 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map