Assay Details
Target Gene Details
Entrez Gene ID: | 64866 |
Gene Name: | CUB domain containing protein 1 |
Gene Aliases: |
CD318, SIMA135, TRASK |
Location: |
Chr.3:45082274-45146422 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCP1 | NM_022842.4 | 7 | 1864 | NP_073753.3 |
| XM_011534024.1 | 7 | 1822 | XP_011532326.1 | |
| XM_017007070.1 | 7 | 1850 | XP_016862559.1 | |
| AF468010.1 | 7 | 1839 | AAO33397.1 | |
| AK023834.1 | 5 | 1442 | BAB14695.1 | |
| AK026187.1 | 4 | 881 | BAB15388.1 | |
| AK026622.1 | 7 | 1839 | BAB15511.1 | |
| AK314441.1 | 7 | 1828 | ||
| AY026461.1 | 7 | 1811 | AAK02058.1 | |
| AY167484.1 | 7 | 1821 | ||
| AY375452.1 | 7 | 1810 | AAR21289.1 | |
| BC069254.1 | 7 | 1817 | AAH69254.1 | |
| KU215672.1 | 6 | 1642 | ||
| KU257676.1 | 6 | 1639 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75990347] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map