Assay Details
Target Gene Details
Entrez Gene ID: | 320 |
Gene Name: | amyloid beta precursor protein binding family A member 1 |
Gene Aliases: |
D9S411E, LIN10, MINT1, X11, X11A, X11ALPHA |
Location: |
Chr.9:69427532-69673012 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBA1 | NM_001163.3 | 13 | 2921 | NP_001154.2 |
| XM_005251968.3 | 12 | 2919 | XP_005252025.1 | |
| XM_011518617.2 | 13 | 2876 | XP_011516919.1 | |
| XM_017014670.1 | 13 | 3165 | XP_016870159.1 | |
| AF029106.1 | 13 | 2843 | AAC05304.1 | |
| AF047347.1 | 13 | 2884 | AAC39766.1 | |
| L04953.1 | 12 | 2258 | AAA61307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29180 | Chr.9:69419809 - 69434109 on Build GRCh38 | Gain |
|
| dgv7669n100 | Chr.9:69413542 - 69434676 on Build GRCh38 | Gain |
|
| esv2761289 | Chr.9:69413554 - 69434688 on Build GRCh38 | Gain |
|
| dgv7668n100 | Chr.9:69381047 - 69564931 on Build GRCh38 | Gain |
|
| nsv951525 | Chr.9:69413985 - 69438984 on Build GRCh38 | Duplication |
|
| dgv7670n100 | Chr.9:69417465 - 69454398 on Build GRCh38 | Gain |
|
| esv2422080 | Chr.9:69414684 - 69434112 on Build GRCh38 | Duplication |
|
| nsv442554 | Chr.9:69420277 - 69434112 on Build GRCh38 | Gain |
|
| nsv818703 | Chr.9:69420931 - 69433561 on Build GRCh38 | Gain |
|
| esv2759694 | Chr.9:69403330 - 69433182 on Build GRCh38 | Gain |
|
| esv3620559 | Chr.9:69416055 - 69439564 on Build GRCh38 | Gain |
|
| esv2761532 | Chr.9:69420273 - 69434688 on Build GRCh38 | Gain |
|
| nsv1038185 | Chr.9:69420283 - 69442388 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map