Assay Details
Target Gene Details
Entrez Gene ID: | 58985 |
Gene Name: | interleukin 22 receptor subunit alpha 1 |
Gene Aliases: |
CRF2-9, IL22R, IL22R1 |
Location: |
Chr.1:24119771-24143285 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL22RA1 | NM_021258.3 | NP_067081.2 | ||
| XM_011541882.1 | XP_011540184.1 | |||
| AF286095.1 | AAG22073.1 | |||
| AK226001.1 | ||||
| AK292204.1 | ||||
| AK304454.1 | ||||
| AK308522.1 | ||||
| AK313971.1 | ||||
| BC029273.1 | AAH29273.1 |
More Information
Set Membership: |
|

Genomic Map