Assay Details
Target Gene Details
Entrez Gene ID: | 220202 |
Gene Name: | atonal bHLH transcription factor 7 |
Gene Aliases: |
Math5, NCRNA, PHPVAR, RNANC, bHLHa13 |
Location: |
Chr.10:68230595-68232113 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOH7 | NM_145178.3 | 1 | 1381 | NP_660161.1 |
| AB593108.1 | 1 | 1381 | ||
| AB593109.1 | 1 | 1381 | ||
| BC032621.1 | 1 | 1388 | AAH32621.1 | |
| H05728.1 | 1 | 154 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526735 | Chr.10:68223370 - 68232096 on Build GRCh38 | Loss |
|
| esv3623705 | Chr.10:68099008 - 68297411 on Build GRCh38 | Gain |
|
| esv3623707 | Chr.10:68214112 - 68304911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map