Assay Details
Target Gene Details
Entrez Gene ID: | 10215 |
Gene Name: | oligodendrocyte lineage transcription factor 2 |
Gene Aliases: |
BHLHB1, OLIGO2, PRKCBP2, RACK17, bHLHe19 |
Location: |
Chr.21:33025908-33029196 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLIG2 | NM_005806.3 | 2 | 1640 | NP_005797.1 |
| XM_005260908.1 | 2 | 1847 | XP_005260965.1 | |
| AF221520.1 | 2 | 1563 | AAF61215.1 | |
| AK091462.1 | 1 | 2387 | ||
| BC036245.1 | 1 | 2387 | AAH36245.1 | |
| BC047511.1 | 2 | 1613 | AAH47511.1 | |
| U48250.1 | 1 | 1493 | AAC72247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520925 | Chr.21:33021637 - 33033627 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map