Assay Details
Target Gene Details
Entrez Gene ID: | 220 |
Gene Name: | aldehyde dehydrogenase 1 family member A3 |
Gene Aliases: |
ALDH1A6, ALDH6, MCOP8, RALDH3 |
Location: |
Chr.15:100879692-100916626 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1A3 | NM_000693.3 | NP_000684.2 | ||
| NM_001293815.1 | NP_001280744.1 | |||
| AK302607.1 | ||||
| AK303441.1 | ||||
| AK312303.1 | ||||
| BC069274.1 | AAH69274.1 | |||
| BX538027.1 | CAD97973.1 | |||
| U07919.1 | AAA79036.1 |
Target Gene Details
Entrez Gene ID: | 101927751 |
Gene Name: | uncharacterized LOC101927751 |
Gene Aliases: |
- |
Location: |
Chr.15:100892343-100919283 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927751 | NR_135827.1 | |||
| AK126286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041172 | Chr.15:100435008 - 101062634 on Build GRCh38 | Gain |
|
| nsv570827 | Chr.15:100884530 - 100924371 on Build GRCh38 | Loss |
|
| nsv469824 | Chr.15:100759595 - 100934123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map