Assay Details
Target Gene Details
Entrez Gene ID: | 5197 |
Gene Name: | platelet factor 4 variant 1 |
Gene Aliases: |
CXCL4L1, CXCL4V1, PF4-ALT, PF4A, SCYB4V1 |
Location: |
Chr.4:73853296-73854484 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PF4V1 | NM_002620.3 | NP_002611.1 | ||
| BC130653.1 | ||||
| BC130657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
| nsv4389 | Chr.4:73824830 - 73856555 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map