Assay Details
Target Gene Details
Entrez Gene ID: | 222894 |
Gene Name: | Fer3-like bHLH transcription factor |
Gene Aliases: |
N-TWIST, NATO3, NTWIST, bHLHa31 |
Location: |
Chr.7:19144782-19145421 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FERD3L | NM_152898.2 | NP_690862.1 | ||
| AF517122.1 | AAN04086.1 | |||
| BC101135.1 | AAI01136.1 | |||
| BC101136.1 | AAI01137.1 | |||
| BC101137.1 | AAI01138.1 | |||
| BC101138.2 | AAI01139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022118 | Chr.7:19138452 - 19478537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map