Assay Details
Target Gene Details
Entrez Gene ID: | 10092 |
Gene Name: | actin related protein 2/3 complex subunit 5 |
Gene Aliases: |
ARC16, dJ127C7.3, p16-Arc |
Location: |
Chr.1:183626193-183635941 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC5 | NM_001270439.1 | NP_001257368.1 | ||
| NM_005717.3 | NP_005708.1 | |||
| AF006088.1 | AAB64193.1 | |||
| AF017807.1 | AAB70561.1 | |||
| AK095424.1 | ||||
| AK129598.1 | ||||
| BC057237.1 | AAH57237.1 | |||
| BI758106.1 | ||||
| CN291383.1 |
More Information
Set Membership: |
|

Genomic Map