Assay Details
Target Gene Details
Entrez Gene ID: | 22950 |
Gene Name: | solute carrier family 4 member 1 adaptor protein |
Gene Aliases: |
HLC3 |
Location: |
Chr.2:27663471-27694980 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A1AP | NM_018158.2 | 1 | 193 | NP_060628.2 |
| AK001486.1 | 1 | 193 | BAA91718.1 | |
| AY028435.1 | 1 | 169 | AAK29177.1 | |
| BC098302.1 | 1 | 188 | AAH98302.1 | |
| BC098358.1 | 1 | 188 | AAH98358.1 | |
| BC099711.1 | 1 | 188 | AAH99711.1 | |
| BC099739.1 | 1 | 186 | AAH99739.1 |
Target Gene Details
Entrez Gene ID: | 9913 |
Gene Name: | SPT7-like STAGA complex gamma subunit |
Gene Aliases: |
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H |
Location: |
Chr.2:27642568-27663840 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT7L | NM_001282729.1 | 1 | 178 | NP_001269658.1 |
| NM_001282730.1 | 1 | 178 | NP_001269659.1 | |
| NM_001282731.1 | 1 | 178 | NP_001269660.1 | |
| NM_001282732.1 | 1 | 178 | NP_001269661.1 | |
| NM_014860.2 | 1 | 178 | NP_055675.1 | |
| XM_005264672.4 | 1 | 162 | XP_005264729.1 | |
| AF224759.1 | AAG47636.1 | |||
| DA001757.1 | ||||
| DA398215.1 | 1 | 180 |
More Information
Set Membership: |
|

Genomic Map