Assay Details
Target Gene Details
Entrez Gene ID: | 84292 |
Gene Name: | WD repeat domain 83 |
Gene Aliases: |
MORG1 |
Location: |
Chr.19:12666804-12675832 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR83 | NM_001099737.2 | NP_001093207.1 | ||
| NR_029375.1 | ||||
| AK074525.1 | ||||
| AK314164.1 | ||||
| Z46189.1 |
Target Gene Details
Entrez Gene ID: | 51398 |
Gene Name: | WD repeat domain 83 opposite strand |
Gene Aliases: |
C19orf56, PTD008 |
Location: |
Chr.19:12668067-12671023 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR83OS | NM_016145.3 | 4 | 642 | NP_057229.1 |
| XM_017026864.1 | 5 | 1636 | XP_016882353.1 | |
| AA694460.1 | 1 | 220 | ||
| AF059620.1 | 4 | 410 | AAG43119.1 | |
| AF078861.1 | 4 | 627 | AAD44493.1 | |
| AF151898.1 | 4 | 626 | AAD34135.1 | |
| AK091030.1 | 4 | 2347 | ||
| BC001192.1 | 4 | 390 | AAH01192.1 | |
| BC042919.1 | 4 | 411 | AAH42919.1 | |
| BC061912.1 | 4 | 411 | AAH61912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
| dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map