Assay Details
Target Gene Details
Entrez Gene ID: | 64403 |
Gene Name: | cadherin 24 |
Gene Aliases: |
CDH11L |
Location: |
Chr.14:23047061-23057619 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH24 | NM_022478.3 | 13 | 2266 | NP_071923.2 |
| NM_144985.3 | 12 | 2152 | NP_659422.2 | |
| AK025342.1 | 5 | 570 | BAB15116.1 | |
| AK289576.1 | 12 | 2152 | ||
| AL137477.1 | 4 | 725 | CAB70758.1 | |
| AY260900.1 | 11 | 1985 | AAP20590.1 | |
| AY260901.1 | 12 | 2099 | AAP20591.1 | |
| AY358199.1 | 12 | 2108 | AAQ88566.1 | |
| BC094810.1 | AAH94810.1 | |||
| BC110867.1 | 5 | 722 | AAI10868.1 |
More Information
Set Membership: |
|

Genomic Map