Assay Details
Target Gene Details
Entrez Gene ID: | 4613 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog |
Gene Aliases: |
MODED, N-myc, NMYC, ODED, bHLHe37 |
Location: |
Chr.2:15940438-15947007 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCN | NM_001293228.1 | 1 | 224 | NP_001280157.1 |
| NM_001293231.1 | 1 | 224 | NP_001280160.1 | |
| NM_001293233.1 | 1 | 224 | NP_001280162.1 | |
| NM_005378.5 | 1 | 224 | NP_005369.2 | |
| AF320053.1 | 1 | 98 | AAG40001.1 | |
| AK298225.1 | 1 | 104 | ||
| AK299495.1 | 1 | 98 | ||
| M55330.1 | 1 | 224 |
Target Gene Details
Entrez Gene ID: | 10408 |
Gene Name: | MYCN opposite strand |
Gene Aliases: |
MYCN-AS1, N-CYM, NCYM, NYCM |
Location: |
Chr.2:15936265-15941723 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCNOS | NR_026766.1 | |||
| NR_110230.1 | ||||
| S49953.1 | AAB24206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1102e212 | Chr.2:15937643 - 15955551 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map