Assay Details
Target Gene Details
Entrez Gene ID: | 55969 |
Gene Name: | chromosome 20 open reading frame 24 |
Gene Aliases: |
PNAS-11, RIP5 |
Location: |
Chr.20:36605734-36612557 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C20orf24 | NM_001199534.1 | 4 | 692 | NP_001186463.1 |
| NM_018840.4 | 4 | 664 | NP_061328.1 | |
| NM_199483.2 | 3 | 534 | NP_955777.1 | |
| NR_026562.3 | 5 | 775 | ||
| AF112213.1 | 4 | 628 | AAF17201.1 | |
| AF274936.1 | 4 | 562 | AAK07515.1 | |
| AF274948.1 | 4 | 477 | AAK07525.1 | |
| BC001871.1 | 4 | 609 | AAH01871.1 | |
| BI223439.1 | ||||
| BQ086248.1 | ||||
| BU189639.1 | 3 | 446 | ||
| BU848918.1 | 4 | 623 | ||
| CD370956.1 | 1 | 465 |
Target Gene Details
Entrez Gene ID: | 100527943 |
Gene Name: | TGIF2-C20orf24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:36574554-36612557 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF2-C20orf24 | NM_001199535.1 | 5 | 640 | NP_001186464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map