Assay Details
Target Gene Details
Entrez Gene ID: | 27125 |
Gene Name: | AF4/FMR2 family member 4 |
Gene Aliases: |
AF5Q31, CHOPS, MCEF |
Location: |
Chr.5:132875379-132963662 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFF4 | NM_014423.3 | 21 | 8244 | NP_055238.1 |
| XM_005271963.4 | 22 | 8308 | XP_005272020.1 | |
| XM_005271964.4 | 15 | 6956 | XP_005272021.1 | |
| XM_006714587.3 | 20 | 8157 | XP_006714650.1 | |
| AF173886.1 | 1 | 255 | AAQ13633.1 | |
| AK126818.1 | 1 | 134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830489 | Chr.5:132815845 - 133000744 on Build GRCh38 | Loss |
|
| esv3606811 | Chr.5:132860904 - 132910818 on Build GRCh38 | Gain |
|
| nsv5003 | Chr.5:132845164 - 132889919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map