Assay Details
Target Gene Details
Entrez Gene ID: | 6510 |
Gene Name: | solute carrier family 1 member 5 |
Gene Aliases: |
AAAT, ASCT2, ATBO, M7V1, M7VS1, R16, RDRC |
Location: |
Chr.19:46759685-46788622 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A5 | NM_001145144.1 | 5 | 349 | NP_001138616.1 |
| NM_001145145.1 | 4 | 539 | NP_001138617.1 | |
| NM_005628.2 | 5 | 1485 | NP_005619.1 | |
| XM_005259167.4 | 5 | 1522 | XP_005259224.1 | |
| AB209738.1 | 5 | 1451 | BAD92975.1 | |
| AF102826.1 | 5 | 886 | AAD09812.1 | |
| AF105230.1 | 5 | 1239 | AAD09814.1 | |
| AF105423.1 | 5 | 1455 | AAD27806.1 | |
| AF334818.1 | 5 | 828 | AAK77026.1 | |
| AK055630.1 | 4 | 645 | ||
| AK125222.1 | 6 | 863 | ||
| AK292690.1 | 5 | 1494 | ||
| AK293433.1 | 5 | 346 | ||
| AK299137.1 | 4 | 486 | ||
| AK301661.1 | 5 | 349 | ||
| AK316546.1 | 4 | 452 | ||
| BC000062.2 | 5 | 1005 | AAH00062.1 | |
| GQ919058.1 | 5 | 865 | ||
| U53347.1 | 5 | 1484 | AAC50629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893223 | Chr.19:46773197 - 46810074 on Build GRCh38 | Gain |
|
| esv1008167 | Chr.19:46777837 - 46789718 on Build GRCh38 | Deletion |
|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
| nsv1065221 | Chr.19:46772775 - 46831220 on Build GRCh38 | Gain |
|
| dgv3608n100 | Chr.19:46757741 - 46865807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map