Assay Details
Target Gene Details
Entrez Gene ID: | 283899 |
Gene Name: | INO80 complex subunit E |
Gene Aliases: |
CCDC95 |
Location: |
Chr.16:29996209-30005794 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80E | NM_001304562.1 | 6 | 693 | NP_001291491.1 |
| NM_001304563.1 | 8 | 900 | NP_001291492.1 | |
| NM_173618.2 | 7 | 810 | NP_775889.1 | |
| NR_130786.1 | 8 | 1102 | ||
| NR_130787.1 | 7 | 1012 | ||
| NR_134855.1 | 11 | 1590 | ||
| XM_011545809.2 | XP_011544111.1 | |||
| AK074033.1 | 1 | 4002 | BAB84859.1 | |
| AK074137.1 | 7 | 4110 | BAB84963.1 | |
| AK075133.1 | 7 | 809 | BAC11424.1 | |
| AK097889.1 | 1 | 1549 | ||
| AK130409.1 | 7 | 801 | ||
| AL525020.3 | 7 | 810 | ||
| AY189289.1 | 10 | 2688 | AAO86733.1 | |
| BC047712.2 | 7 | 771 | AAH47712.1 | |
| BU528528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map