Assay Details
Target Gene Details
Entrez Gene ID: | 8405 |
Gene Name: | speckle type BTB/POZ protein |
Gene Aliases: |
BTBD32, TEF2 |
Location: |
Chr.17:49598884-49678163 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOP | NM_001007226.1 | 12 | 2579 | NP_001007227.1 |
| NM_001007227.1 | 11 | 2457 | NP_001007228.1 | |
| NM_001007228.1 | 10 | 2405 | NP_001007229.1 | |
| NM_001007229.1 | 11 | 2456 | NP_001007230.1 | |
| NM_001007230.1 | 12 | 2508 | NP_001007231.1 | |
| NM_003563.3 | 11 | 2527 | NP_003554.1 | |
| BQ226031.1 | 1 | 346 |
More Information
Set Membership: |
|

Genomic Map