Assay Details
Target Gene Details
Entrez Gene ID: | 498 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle |
Gene Aliases: |
ATP5A, ATP5AL2, ATPM, COXPD22, HEL-S-123m, MC5DN4, MOM2, OMR, ORM, hATP1 |
Location: |
Chr.18:46084144-46104233 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5A1 | NM_001001935.2 | NP_001001935.1 | ||
| NM_001001937.1 | NP_001001937.1 | |||
| NM_001257334.1 | NP_001244263.1 | |||
| NM_001257335.1 | NP_001244264.1 | |||
| NM_004046.5 | NP_004037.1 | |||
| XM_017025789.1 | XP_016881278.1 | |||
| AK092735.1 | ||||
| AK098765.1 | ||||
| AK129739.1 | ||||
| AK129804.1 | ||||
| AK129900.1 | ||||
| AK289457.1 | ||||
| AK302272.1 | ||||
| AW205533.1 | ||||
| AW207528.1 | ||||
| BC003119.1 | AAH03119.1 | |||
| BC007299.1 | AAH07299.1 | |||
| BC008028.2 | AAH08028.2 | |||
| BC011384.1 | AAH11384.1 | |||
| BC016046.1 | AAH16046.1 | |||
| BC019310.1 | AAH19310.1 | |||
| BC039135.1 | ||||
| BC064562.1 | AAH64562.1 | |||
| BC067385.1 | AAH67385.1 | |||
| BT007209.1 | AAP35873.1 | |||
| D14710.1 | BAA03531.1 | |||
| FJ224309.1 | ||||
| X59066.1 | CAA41789.1 | |||
| X65460.1 | CAA46452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3642420 | Chr.18:45996630 - 46208987 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map